Learn More
FGF21, Mouse, Clone: 1A8, Abnova™
Description
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this growth factor has not yet been determined. [provided by RefSeq
Specifications
Specifications
| Antigen | FGF21 |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 1A8 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full length recombinant FGF21. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | FGF21 |
| Gene Accession No. | BC018404 |
| Gene Symbols | FGF21 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.