Learn More
fibroblast growth factor 19, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human FGF19 protein.
Brand: Abnova H00009965-B01.50uL
Description
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes including embryonic development cell growth, morphogenesis, tissue repair, tumor growth and invasion. This growth factor is a high affinity, heparin dependent ligand for FGFR4. Expression of this gene was detected only in fetal but not adult brain tissue. Synergistic interaction of the chick homolog and Wnt-8c has been shown to be required for initiation of inner ear development. [provided by RefSeq
Sequence: MRSGCVVVHVWILAGLWLAVAGRPLAFSDAGPHVHYGWGDPIRLRHLYTSGPHGLSSCFLRIRADGVVDCARGQSAHSLLEIKAVALRTVAIKGVHSVRYLCMGADGKMQGLLQYSEEDCAFEEEIRPDGYNVYRSEKHRLPVSLSSAKQRQLYKNRGFLPLSHFLPMLPMVPEEPEDLRGHLESDMFSSPLETDSMDPFGLVTGLEAVRSPSFEKSpecifications
| fibroblast growth factor 19 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human FGF19 protein. | |
| FGF19 | |
| FGF19 | |
| FGF19 (AAH17664, 1 a.a. ∼ 216 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 9965 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| No additive | |
| BC017664 | |
| Mouse | |
| 50 μL | |
| Neurobiology | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.