missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-FGD3 Ab-2 Matched Antibody Pair, Abnova™
Mached Pair Antibody
Brand: Abnova H00089846-AP21.1Set
Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-FGD3 (100μg); Detection antibody: mouse purified polyclonal anti-FGD3 (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
Host Species: Rabbit, Mouse
Cross Reactivity: Human
ELISA Pair
Specifications
| FGD3 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human FGD3. | |
| FLJ00004/MGC117260/ZFYVE5 | |
| Mouse, Rabbit | |
| 1 Set | |
| 89846 | |
| Human |
| ELISA | |
| Unconjugated | |
| FGD3 | |
| FGD3 | |
| Affinity Purified | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-FGD3 (100 μg) 2. Detection antibody: mouse purified polyclonal anti-FGD3 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido