Learn More
Fcμ Receptor Mouse anti-Human, Biotin, Clone: HM14-1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 563018
Description
The HM14-1 monoclonal antibody specifically binds to the FAIM3 -encoded high affinity receptor for IgM (also known as, Fcµ Receptor, FcμR, Fc mu receptor, Immunoglobulin mu Fc receptor, FcmR, IgM Fc receptor). The FcμR is a type 1 transmembrane glycoprotei n that contains an extracellular Ig-like domain with sequence similarities to Fcα/μR and pIgR. It is likewise known as Fas apoptotic inhibitory molecule 3, TOSO or Regulator of Fas-induced apoptosis Toso based on its capacity to counter apoptosis mediated by Fas/CD95 (or TNF) in some experimental model systems. FcμRs are expressed by B cells, CD4 + T cells, CD8+ T cells, and NK cells and are highly expressed on Chronic Lymphocytic Leukemia cells. FcμRs may play roles in cellular activation, antigen presentation and IgM-dependent cell-mediated cytotoxicity. The relatively long cytoplasmic tail of FcμR contains several conserved tyrosine and serine residues. These get phosphorylated after FcμR ligation with Ig M immune complexes and may initiate signaling responses. Lymphocyte FcμRs can mediate the internalization of IgM-bound antigens including microbial and viral antigens. This in turn may cause cellular activation due to subsequent antigen processing and exposure of activating ligands to intracellular receptors including Toll-like receptors. The HM7-10 antibody, a related FcμR-specific antibody, reportedly recognizes an epitope near the IgM ligand binding site of the FcμR whereas the HM14-1 antibody does not.
Specifications
| Fcμ Receptor | |
| Monoclonal | |
| 0.5mg/mL | |
| FcμR; FcmR; Fc mu receptor; IgM Fc receptor; FAIM3; TOSO | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| HM14-1 | |
| Biotin | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human IgM Fc Receptor Transfected Cell Line | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.