Learn More
F-box protein 6, Mouse, Clone: 7D1, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant FBXO6.
Brand: Abnova H00026270-M02A.200uL
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class, and its C-terminal region is highly similar to that of rat NFB42 (neural F Box 42kDa) which may be involved in the control of the cell cycle. [provided by RefSeq]
Sequence: MDAPHSKAALDSINELPENILLELFTHVPARQLLLNCRLVCSLWRDLIDLMTLWKRKCLREGFITKDWDQPVADWKIFYFLRSLHRNLLRNPCAEEDMFAWQIDFNGGDRWKVESLPGAHGTDFPDPKVKKYFVTSYEMCLKSQLVDLVAEGYWEELLDTFRPDIVVKDWFAARADCGCTYQLKVQLASADYFVLASFEPPPVTIQQWNNATWTEVSYTFSDYPRGVRYILFQHGGRDTQYWAGWYGPRVTNSSIVVSPKMTRNQASSEAQPGQKHGQEEAAQSPYRAVVQIFSpecifications
| F-box protein 6 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC020880 | |
| FBXO6 | |
| FBXO6 (AAH20880.1, 1 a.a. ∼ 293 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| Yes | |
| 26270 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Immunoprecipitation, Western Blot | |
| 7D1 | |
| Mouse monoclonal antibody raised against a full-length recombinant FBXO6. | |
| FBXO6 | |
| FBG2/FBS2/FBX6/Fbx6b | |
| Mouse | |
| 200 μL | |
| Ubiquitin/Proteasome | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.