missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA526203
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology.
FBXO3 encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs.
Specifications
| FBXO3 | |
| Polyclonal | |
| FBXO3 | |
| FBA, FBX3, F-box only protein 3, F-box protein FBX3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26273 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Q9UK99 | |
| FBXO3 | |
| KLH conjugated synthetic peptide between 395-422 amino acids from the C-terminal region of human FBXO3 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction