Learn More
FBXL3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522133
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Molt-4, C2C12. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (95%), Sheep (100%), Rhesus Monkey (100%), Chimpanzee (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FBXL3 Polyclonal antibody specifically detects FBXL3 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
FBXL3 | |
Polyclonal | |
Unconjugated | |
FBXL3 | |
Afh; AU041772; AW212966; FBK; FBL3; Fbl3a; F-box and leucine rich repeat protein 3; F-box and leucine-rich repeat protein 3; F-box and leucine-rich repeat protein 3A; F-box protein Fbl3a; F-box/LRR-repeat protein 3; F-box/LRR-repeat protein 3A; FBXL3; Fbxl3a; Ovtm; Play68; Protein after-hours; protein overtime | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26224, 50789 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.41 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q8C4V4, Q9UKT7 | |
FBXL3 | |
Recombinant fragment corresponding to a region within amino acids 14 and 309 of FBXL3 (Uniprot ID#Q9UKT7) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |