missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM82B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542837
This item is not returnable.
View return policy
Description
Peptide sequence: MALAARLWRL LPFRRGAAPG SRLPAGTSGS RGHCGPCRFR GFEVMGNPGT Sequence homology: Human: 100%
FAM82B Polyclonal antibody specifically detects FAM82B in Human samples. It is validated for Western Blot.Specifications
FAM82B | |
Polyclonal | |
Unconjugated | |
RMDN1 | |
CGI-90; Fam82b; family with sequence similarity 82, member B; hRMD-1; microtubule-associated protein; Protein FAM82B; regulator of microtubule dynamics 1; regulator of microtubule dynamics protein 1; RMD1; RMD-1; RMDN1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51115 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96DB5 | |
RMDN1 | |
Synthetic peptide directed towards the N-terminal of human FAM82B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |