Learn More
FADD Mouse anti-Human, Unlabeled, Clone: 1, BD
Description
During apoptosis, cells exhibit morphological signs of the death process: cell shrinkage, membrane blebbing, and chromatin condensation. The role of the cell surface cytokine receptor, Fas (Apo-1, CD95), in apoptosis has been well characterized. The tumor necrosis factor receptor (TNF-R) can trigger cell death, as well as various other responses. Data suggested that Fas and TNF-R affect a common target in the cell death pathway. This target has been identified as FADD, a novel protein that contains a death domain homologous to the death domains of Fas and TNF-R1. FADD specifically binds to Fas, an association mediated by their homologous death domains. Overexpression of FADD induces apoptosis that is inhibited by CrmA, a poxvirus protein that blocks both Fas- and TNF-induced cell death. Thus, FADD is a central element of the Fas-mediated cell death pathway.
This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
Specifications
| Antigen | FADD |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 1 |
| Concentration | 250μg/mL |
| Conjugate | Unconjugated |
| Formulation | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.