missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Factor V Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA143041
This item is not returnable.
View return policy
Description
PA1-43041 detects Factor V in human samples. PA1-43041 has successfully been used in Western blot and ELISA applications. PA1-43041 was derived from human factor V.
Coagulation factor V (or Proaccelerin) is an essential factor of the blood coagulation cascade. This factor circulates in plasma, and is converted to the active form by the release of the activation peptide by thrombin during coagulation. This generates a heavy chain and a light chain which are held together by calcium ions. The active factor V is a cofactor that participates with activated coagulation factor X to activate prothrombin to thrombin. Defects in the Factor V gene can result various diseases including Factor V deficiency, thrombophilia due to activated protein C resistance, Budd-Chiari syndrome, Ischemic stroke, and recurrent pregnancy loss.
Specifications
| Factor V | |
| Polyclonal | |
| Unconjugated | |
| F5 | |
| Ac2-120; Activated protein C cofactor; AI173222; Cf5; Cf-5; coagulation factor V; coagulation factor V (proaccelerin, labile factor); Coagulation factor V heavy chain; coagulation factor V jinjiang A2 domain; Coagulation factor V light chain; F5; Factor 5; factor V; factor V Leiden; Factor5; FVL; PCCF; Proaccelerin, labile factor; RPRGL1; THPH2 | |
| Sheep | |
| SDS-PAGE | |
| RUO | |
| 2153 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 9.9 mg/mL | |
| 50% water with 50% glycerol and no preservative | |
| P12259 | |
| F5 | |
| Human Factor V. | |
| 1 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction