Learn More
FABP7, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant FABP7.
Brand: Abnova H00002173-A01.50uL
Description
The protein encoded by this gene is a brain fatty acid binding protein. Fatty acid binding proteins (FABPs) are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABPs are thought to play roles in fatty acid uptake, transport, and metabolism. [provided by RefSeq
Sequence: MVEAFCATWKLTNSQNFDEYMKALGVGFATRQVGNVTKPTVIISQEGDKVVIRTLSTFKNTEISFQLGEEFDETTADDRNCKSVVSLDGDKLVHIQKWDGKETNFVREIKDGKMVMTLTFGDVVAVRHYEKASpecifications
| FABP7 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length recombinant FABP7. | |
| FABP7 | |
| B-FABP/BLBP/DKFZp547J2313/FABPB/MRG | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC012299 | |
| FABP7 | |
| FABP7 (AAH12299, 1 a.a. ∼ 132 a.a) full-length recombinant protein with GST tag. | |
| RUO | |
| 2173 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.