Learn More
FABP4, Mouse, Clone: 3G1, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant FABP4.
Brand: Abnova H00002167-M05.100ug
Description
FABP4 encodes the fatty acid binding protein found in adipocytes. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism. [provided by RefSeq
Sequence: MCDAFVGTWKLVSSENFDDYMKEVGVGFATRKVAGMAKPNMIISVNGDVITIKSESTFKNTEISFILGQEFDEVTADDRKVKSTITLDGGVLVHVQKWDGKSTTIKRKREDDKLVVECVMKGVTSTRVYERASpecifications
| FABP4 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC003672 | |
| FABP4 | |
| FABP4 (AAH03672, 1 a.a. ∼ 132 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 3G1 | |
| Mouse monoclonal antibody raised against a full length recombinant FABP4. | |
| FABP4 | |
| A-FABP/aP2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2167 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.