Learn More
fatty acid binding protein 3, muscle and heart (mammary-derived growth inhibitor), Mouse, Clone: 4F6-1D6, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant FABP3.
Brand: Abnova H00002170-M01.100ug
Description
The intracellular fatty acid-binding proteins (FABPs) belongs to a multigene family. FABPs are divided into at least three distinct types, namely the hepatic-, intestinal- and cardiac-type. They form 14-15kDa proteins and are thought to participate in the uptake, intracellular metabolism and/or transport of long-chain fatty acids. They may also be responsible in the modulation of cell growth and proliferation. Fatty acid-binding protein 3 gene contains four exons and its function is to arrest growth of mammary epithelial cells. This gene is a candidate tumor suppressor gene for human breast cancer. [provided by RefSeq
Sequence: MVDAFLGTWKLVDSKNFDDYMKSLGVGFATRQVASMTKPTTIIEKNGDILTLRTHSTFKNTEISFKLGVEFDETTADDRKVKSIVTLDGGKLVHLQKWDGQETTLVRELIDGKLILTLTHGTAVCTRTYEKEASpecifications
| fatty acid binding protein 3, muscle and heart (mammary-derived growth inhibitor) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC007021 | |
| FABP3 | |
| FABP3 (AAH07021, 1 a.a. ∼ 133 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Cell Cycle | |
| 2170 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA, KnockDown, Western Blot | |
| 4F6-1D6 | |
| Mouse monoclonal antibody raised against a full length recombinant FABP3. | |
| FABP3 | |
| FABP11/H-FABP/MDGI/O-FABP | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.