missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PAR3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA533529

Product Code. 15334681

  • £441.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Percent identity with other species by BLAST analysis: Human, Gorilla, Marmoset (100%) Monkey, Bovine, Panda (95%) Gibbon, Dog, Elephant, Pig (89%) Rat, Hamster (84%).

This polyclonal antibody was raised in Rabbit against F2RL2 / PAR3. This antibody is unconjugated. This antibody is useful for testing in the following assays: ICC, IHC-P. The specificity of this antibody has been described as follows: Human F2RL2. BLAST analysis of the peptide immunogen showed no homology with other human proteins.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PAR3
Polyclonal
Unconjugated
F2RL2
coagulation factor II (thrombin) receptor-like 2; coagulation factor II (thrombin) receptor-like 2 precursor; coagulation factor II receptor-like 2; Coagulation factor II receptor-like 2 (protease-actovated receptor 3); coagulation factor II thrombin receptor like 2; F2RL2; PAR3; PAR-3; protease-activated receptor 3; proteinase-activated receptor 3; proteinase-activated receptor-3; thrombin receptor-like 2
Rabbit
Antigen Affinity Chromatography
RUO
2151
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
O00254
F2RL2
Synthetic 19 amino acid peptide from internal region of human F2RL2.
50 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.