missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EZH2 Mouse, Unlabeled, Clone: 11, BD
Mouse Monoclonal Antibody
£181.00 - £314.49
Specifications
| Antigen | EZH2 |
|---|---|
| Clone | 11 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15846259
|
BD Biosciences
612666 |
50 μg |
£181.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15245123
|
BD Biosciences
612667 |
150 μg |
£328.00 £314.49 / Each Save £13.51 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The 11/EZH2 monoclonal antibody specifically binds to the methyltransferase, EZH2 (Enhancer of Zeste Homolog 2). EZH2 is a human homologue of Drosophila's Enhancer of zeste gene, an important regulator of homeobox gene expression. The EZH2 protein has a predicted molecular weight of ∼85 kDa. EZH2 is a member of the Polycomb group (PcG) of proteins that are essential for the maintenance, but not initiation, of the transcriptionally repressed state of certain developmental genes. PcG proteins are a structurally diverse group of proteins with conserved functions from fly to human cells. PcG family proteins form multimeric complexes that regulate the expression of genes involved in cell cycle, DNA repair and differentiation. Specifically, EZH2 is a core enzymatic component of PRC2 (polycomb repressive complex 2). EZH2 is expressed in some lymph node follicular T cells and B cells. Thymocytes differentially express EZH2 at various stages during T-cell maturation. EZH2 interacts with multiple signaling proteins, including Vav, that are involved in lymphocyte development and activation. It is highly expressed in a variety of tumors including lymphomas as well as breast and prostate cancers. EZH2 is important in the self renewal and proliferation of numerous stem cell types including fetal hematopoietic stem cells, muscle satellite cells, hepatic stem/progenitor cells, neural stem cells, basal cell progenitors in the developing epidermis, embryonic stem cells, and some cancer stem cells.Specifications
| EZH2 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 11 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Chicken, Human, Mouse, Rat | |
| Human EZH2 recombinant protein aa. 156-256 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title