missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETV4, Mouse, Clone: 1A3-1D3, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant ETV4.
Brand: Abnova H00002118-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: MYLHTEGFSGPSPGDGAMGYGYEKPLRPFPDDVCVAPEKFEGDIKQEGVGAFREGPPYQRRGALQLWQFLVALLDDPTNAHFIAWTGRGMEFKLIEPEEVARLWGIQKNRPAMNYDKLSRSLRYYYEKGIMQKVAGERYVYKFVCEPEALFSLAFPDNQRPALKAEFDRPVSEEDTVPLSHLDESPAYLPELAGPAQPFGPKGGYSYSpecifications
| ETV4 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC007242 | |
| ETV4 | |
| ETV4 (AAH07242, 1 a.a. ∼ 207 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 1A3-1D3 | |
| Mouse monoclonal antibody raised against a full-length recombinant ETV4. | |
| ETV4 | |
| E1A-F/E1AF/PEA3/PEAS3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2118 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction