missing translation for 'onlineSavingsMsg'
Learn More

Estrogen Receptor alpha Monoclonal Antibody (1D5), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA513191

Product Code. 11304803

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Staining of formalin-fixed, paraffin-embedded tissues requires boiling tissues in 10mM citrate buffer (pH 6.0) for 10-20 min, followed by cooling at room temperature for 20 min. A recommended positive control is MCF-7 cells. For optimal IF staining, it is recommended to fix and permeabilize cells with methanol.

Estrogen Receptor alpha Monoclonal antibody specifically detects Estrogen Receptor alpha in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Estrogen Receptor alpha
Monoclonal
0.8 mg/mL
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
P03372
ESR1
Recombinant human estrogen receptor protein
500 μL
Primary
Human
Antibody
IgG1 κ
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
1D5
Unconjugated
ESR1
DKFZp686N23123; ER; Er alpha; ER36; ERa; ERalpha; ER-alpha; ERalpha protein; ESR; esr 1; Esr1; esr-1; ESRA; Estr; Estra; Estradiol receptor; estrogen nuclear receptor alpha; Estrogen receptor; estrogen receptor 1; estrogen receptor 1 (alpha); estrogen receptor alpha; estrogen receptor alpha c-terminus splice variant 1-2; estrogen receptor alpha E1-E2-1-2; estrogen receptor alpha E1-N2-E2-1-2; estrogen receptor alpha splice variant, CTERP-1; estrogen receptor alpha splice variant, ERalphaDup5; estrogen receptor alpha splice variant, ERalphai45a; estrogen receptor alpha splice variant, ERalphai45bL; estrogen receptor alpha splice variant, ERalphai45bS; estrogen receptor alpha splice variant, ERalphai45c; estrogen receptor alpha splice variant, ERalphai56; estrogen receptor alpha splice variant, ERalphai67; estrogen receptor alpha variant delta 4; estrogen receptor protein; estrogen receptor, alpha; ESTRR; hER-alpha36; I79_001166; Nr3a1; nuclear receptor; nuclear receptor subfamily 3 group A member 1; RNESTROR; RP1-130E4.1; unnamed protein product
Mouse
Protein G
RUO
2099
4° C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.