missing translation for 'onlineSavingsMsg'
Learn More

anti-Erythroid Cell Marker/TER-119 Antigen PE-Cy7™, Clone: TER-119, Invitrogen™

Erythroid Cell Marker/TER-119 Antigen Monoclonal antibody A18452 from Invitrogen specifically detects Erythroid Cell Marker/TER-119 Antigen in Mouse samples. It is validated for Flow Cytometry.

Brand:  Invitrogen A18452

Product Code. 15384216

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Erythroid Cell Marker/TER-119 Antigen Monoclonal antibody A18452 from Invitrogen specifically detects Erythroid Cell Marker/TER-119 Antigen in Mouse samples. It is validated for Flow Cytometry.

TER-119 is a lineage marker for erythroid cells from early proerythroblast to mature erythrocyte stages in adult blood, spleen, and bone marrow. It is also present in yolk sac, and fetal and newborn liver. The TER-119 antigen is not expressed by cells of earlier erythroid development at BFU-e (blast-forming unit erythroid) stage or CFU-e (colony-forming unit erythroid) stage.
Host Species: Rat
Cross Reactivity: Mouse
Isotype: IgG2b, κ
Clone: TER-119
Immunogen: Mouse Erythroid Cells.
Formulation: 50μg Purified Antibody (0.2mg/ml) in proprietary buffer, with 0.09% sodium azide
This product detects Erythroid Cell Marker/TER-119 Antigen in Mouse samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is Mouse Erythroid Cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Erythroid Cell Marker/TER-119 Antigen
Monoclonal
PE-Cyanine7
Rat
50 μg
Mouse
Purified
Flow Cytometry
TER-119
50μg Purified Antibody (0.2mg/ml) in proprietary buffer, with 0.09% sodium azide
Mouse Erythroid Cells.
Primary
Store at 4° C
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.