Learn More
ERK1/2 (pT202/pY204) Mouse, PE-Cy7, Clone: 20A, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560116
Description
The members of the Mitogen-Activated Protein Kinase (MAPK) family are components of a key signal transduction cascade that links events at the cell surface to responses in the nucleus. The signaling cascade is found in species as varied as yeast and humans, with many of the proteins being well conserved. In mammals the most widely studied members of the cascade are the Extracellular signal-Regulated Kinases, ERK1 (p44 MAPK) and ERK2 (p42 MAPK). ERK1 and ERK2 share 85% homology and are activated by extracellular signals such as growth factors, hormones, and phorbol esters. Activation occurs through a series of phosphorylations by kinases activating other kinases and eventually leading to phosphorylation of the ERKs. Growth factor stimulation leads to activation of Ras and Raf, leading to phosphorylation of MEK1 (MAPK/ERK kinase) which, in turn, activates the ERKs via dual phosphorylation. Once activated, the ERKs phosphorylate other cytoplasmic signalling molecules, cell-surface receptors, microtubule-associated proteins, and transcription factors in the nucleus. Thus, the active ERK has myriad downstream effectors that implicate it in the control of cell proliferation and differentiation, as well as regulation of the cytoskeleton. Furthermore, studies have shown that elevated ERK activity is associated with some cancers. The 20A monoclonal antibody recognizes the phosphorylated threonine 202 and tyrosine 204 (pT202/pY204) of human ERK1 and pT184/pY186 of human ERK2. The orthologous phosphorylation sites in murine ERK1 and ERK2 are T203/Y205 and T183/Y185.
Host Species: Mouse
Clone: 20A
Isotype: IgG1
Species Reactivity: Human
Immunogen: Phosphorylated Rat ERK1 Peptide
Intracellular Staining
Specifications
| ERK1/2 (pT202/pY204) | |
| Monoclonal | |
| PE-Cyanine7 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Phosphorylated Rat ERK1 Peptide | |
| 50 Tests | |
| Cell Biology | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| 20A | |
| p44/42 MAPK; Extracellular signal-Regulated Kinase 1/2 (pT202/Y204) | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.