missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB3/Her3 Mouse anti-Human, Clone: 66223, R&D Systems™
Mouse Monoclonal Antibody has been used in 12 publications
POA
Specifications
Antigen | ErbB3/Her3 |
---|---|
Clone | 66223 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human ErbB3/Her3 Ser20-Thr643 Accession # P21860 |
Host Species | Mouse |
Primary or Secondary | Primary |
Description
ErbB3/Her3 Monoclonal specifically detects ErbB3/Her3 in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
ErbB3/Her3 | |
Mouse myeloma cell line NS0-derived recombinant human ErbB3/Her3 Ser20-Thr643 Accession # P21860 | |
Primary | |
Human | |
Unconjugated | |
2065 | |
Purified | |
Flow Cytometry, Neutralization, CyTOF, ELISA | |
RUO |
66223 | |
Mouse | |
Monoclonal | |
IgG1 | |
c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian) | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only