Learn More
Goat anti-Equine IgM Secondary Antibody, HRP, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA184647
Code : LD
Additional Details : Weight : 0.01000kg
Description
Goat anti horse IgM polyclonal antibody recognizes equine IgM.No cross-reactivity with other equine immunoglobulin classes is seen in immunoelectrophoresis. This antibody may cross-react with IgM from other species.
Equine IgM Polyclonal antibody specifically detects Equine IgM in Equine samples. It is validated for ELISA.Specifications
Equine IgM | |
Polyclonal | |
HRP | |
Goat | |
Affinity chromatography | |
RUO | |
Equine | |
Liquid |
ELISA | |
1 mg/mL | |
PBS with 0.2% BSA and 0.05% ProClin 300 | |
Purified equine IgM. | |
1 mg | |
Secondary | |
4° C, do not freeze | |
IgG |