missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EPS8L2 Polyclonal Antibody

Description
Recommended positive controls: A431, HepG2. Predicted reactivity: Mouse (82%), Rat (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene has been shown to be involved in the regulation of cell proliferation, and in the crosstalk between the adiponectin signalling and insulin signalling pathways. The encoded protein binds many other proteins, including RAB5A, DCC, AKT2, PIK3CA, adiponectin receptors, and proteins of the NuRD/MeCP1 complex. This protein is found associated with endosomal membranes, but can be released by EGF and translocated to the nucleus.
Specifications
Specifications
| Antigen | EPS8L2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | EPS8L2 |
| Gene Accession No. | Q9H6S3 |
| Gene Alias | AI042819; AW545405; Epidermal growth factor receptor kinase substrate 8-like protein 2; epidermal growth factor receptor pathway substrate 8-like protein 2; epidermal growth factor receptor pathway substrate 8-related protein 2; EPS8 like 2; EPS8 related protein 2; Eps8l2; EPS8-like 2; EPS8-like protein 2; EPS8R2; EPS8-related protein 2; PP13181 |
| Gene Symbols | EPS8L2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?