Learn More
EPOR, Mouse, Clone: 3D10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant EPOR.
Brand: Abnova H00002057-M01.100ug
Beschreibung
The erythropoietin receptor is a member of the cytokine receptor family. Upon erythropoietin binding, the erythropoietin receptor activates Jak2 tyrosine kinase which activates different intracellular pathways including: Ras/MAP kinase, phosphatidylinositol 3-kinase and STAT transcription factors. The stimulated erythropoietin receptor appears to have a role in erythroid cell survival. Defects in the erythropoietin receptor may produce erythroleukemia and familial erythrocytosis. [provided by RefSeq
Sequence: PDPKFESKAALLAARGPEELLCFTERLEDLVCFWEEAASAGVGPGNYSFSYQLEDEPWKLCRLHQAPTARGAVRFWCSLPTADTSSFVPLELRVTAASGASpezifikation
| EPOR | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_000121 | |
| EPOR | |
| EPOR (NP_000112, 31 a.a. ∼ 130 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Immunoprecipitation, Western Blot | |
| 3D10 | |
| Mouse monoclonal antibody raised against a partial recombinant EPOR. | |
| EPOR | |
| MGC138358 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2057 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.