Learn More
epilepsy, progressive myoclonus type 2A, Lafora disease (laforin), Mouse, Clone: 6C6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant EPM2A.
Brand: Abnova H00007957-M02.100ug
Description
This gene encodes a dual-specificity phosphatase that associates with polyribosomes. The encoded protein may be involved in the regulation of glycogen metabolism. Mutations in this gene have been associated with myoclonic epilepsy of Lafora. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Sequence: GNGPHHDRCCTYNENNLVDGVYCLPIGHWIEATGHTNEMKHTTDFYFNIAGHQAMHYSRILPNIWLGSCPRQVEHVTIKLKHELGITAVMNFQTEWDIVSpecifications
| epilepsy, progressive myoclonus type 2A, Lafora disease (laforin) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001018041 | |
| EPM2A | |
| EPM2A (NP_001018051, 101 a.a. ∼ 199 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody |
| ELISA, Western Blot | |
| 6C6 | |
| Mouse monoclonal antibody raised against a partial recombinant EPM2A. | |
| EPM2A | |
| EPM2/MELF | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 7957 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.