Learn More
EPHA5, Mouse, Clone: 6F4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant EPHA5.
Brand: Abnova H00002044-M03.100ug
Description
This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Two transcript variants encoding different isoforms have been found for this gene.
Sequence: PSVVRHLAVFPDTITGADSSQLLEVSGSCVNHSVTDEPPKMHCSAEGEWLVPIGKCMCKAGYEEKNGTCQVCRPGFFKASPHIQSCGKCPPHSYTHEEASSpecifications
| EPHA5 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_004439 | |
| EPHA5 | |
| EPHA5 (NP_004430, 234 a.a. ∼ 333 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 6F4 | |
| Mouse monoclonal antibody raised against a partial recombinant EPHA5. | |
| EPHA5 | |
| CEK7/EHK1/HEK7/TYRO4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2044 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.