Learn More
E1A binding protein p300, Mouse, Clone: 1D2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant EP300.
Brand: Abnova H00002033-M02.50ug
Description
This gene encodes the adenovirus E1A-associated cellular p300 transcriptional co-activator protein. It functions as histone acetyltransferase that regulates transcription via chromatin remodeling and is important in the processes of cell proliferation and differentiation. It mediates cAMP-gene regulation by binding specifically to phosphorylated CREB protein. This gene has also been identified as a co-activator of HIF1A (hypoxia-inducible factor 1 alpha), and thus plays a role in the stimulation of hypoxia-induced genes such as VEGF. Defects in this gene are a cause of Rubinstein-Taybi syndrome and may also play a role in epithelial cancer. [provided by RefSeq
Sequence: PPLQHHGQLAQPGALNPPMGYGPRMQQPSNQGQFLPQTQFPSQGMNVTNIPLAPSSGQAPVSQAQMSSSSCPVNSPIMPPGSQGSHIHCPQLPQPALHQNSpecifications
| E1A binding protein p300 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001429 | |
| EP300 | |
| EP300 (NP_001420, 731 a.a. ∼ 830 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 50 μg | |
| Apoptosis | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 1D2 | |
| Mouse monoclonal antibody raised against a partial recombinant EP300. | |
| EP300 | |
| KAT3B/p300 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 2033 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.