Learn More
eNOS (pS633) Mouse anti-Human, Unlabeled, Clone: 37, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 612665
Description
Nitirc oxide synthase (NOS), a cell type specific enzyme, catalyzes the synthesis of nitric oxide (NO). NO is a short-lived radical that transmits signals involved in vasorelaxation, neurotransmission, and cytoxicity. In neurons and endothelial cells, constitutive NOS (cNOS) is activated by agonists that increase intracellular Ca2+; levels and enhance calmodulin binding. Neuronal NOS (nNOS) and endothelial NOS (eNOS) have recognition sites for NADPH, FAD, FMN, and calmodulin. eNOS has a unique N-myristylation consensus sequence that may explain its membrane localization. Various protein kinases have been implicated in regulation of eNOS activity, including AMPK, PKA, PKB/Akt, PKC, and CaM Kinase II. During VEGF stimulation, eNOS is transiently phosphorylated at Ser-1177 by PKB/Akt and dephosphorylated at Thr-495. At later time points, VEGF stimulation leads to an increase in Thr-495 phosphorylation mediated by PKC and a decrease in Ser-1177 phosphorylation. In addition, Ser-633 amd Ser-1177 are phosphorylated by PKA and PKG in vitro. Thus, eNOS activity may be regulated through complex phosphorylation events mediated by multiple kinases at various phosphorylation sites.
Western Blotting
Specifications
| eNOS (pS633) | |
| Monoclonal | |
| 250μg/mL | |
| Nitric Oxide Synthase | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
| Western Blot | |
| 37 | |
| Unconjugated | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Phosphorylated Human eNOS (pS633) Peptide | |
| 150 μg | |
| Cell Biology | |
| Human | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.