missing translation for 'onlineSavingsMsg'
Learn More

eNOS (pS633) Mouse anti-Human, Unlabeled, Clone: 37, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 612664

 View more versions of this product

Product Code. 15826259

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Nitirc oxide synthase (NOS), a cell type specific enzyme, catalyzes the synthesis of nitric oxide (NO). NO is a short-lived radical that transmits signals involved in vasorelaxation, neurotransmission, and cytoxicity. In neurons and endothelial cells, constitutive NOS (cNOS) is activated by agonists that increase intracellular Ca2+; levels and enhance calmodulin binding. Neuronal NOS (nNOS) and endothelial NOS (eNOS) have recognition sites for NADPH, FAD, FMN, and calmodulin. eNOS has a unique N-myristylation consensus sequence that may explain its membrane localization. Various protein kinases have been implicated in regulation of eNOS activity, including AMPK, PKA, PKB/Akt, PKC, and CaM Kinase II. During VEGF stimulation, eNOS is transiently phosphorylated at Ser-1177 by PKB/Akt and dephosphorylated at Thr-495. At later time points, VEGF stimulation leads to an increase in Thr-495 phosphorylation mediated by PKC and a decrease in Ser-1177 phosphorylation. In addition, Ser-633 amd Ser-1177 are phosphorylated by PKA and PKG in vitro. Thus, eNOS activity may be regulated through complex phosphorylation events mediated by multiple kinases at various phosphorylation sites.

Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

eNOS (pS633)
Monoclonal
250μg/mL
Nitric Oxide Synthase
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
37
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Phosphorylated Human eNOS (pS633) Peptide
50 μg
Cell Biology
Human
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.