Learn More
eNOS Monoclonal Antibody (6H2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515559
Code : A0
Additional Details : Weight : 0.01000kg
Description
MA5-15559 targets eNOS in indirect ELISA, IHC applications and shows reactivity with Human samples. The MA5-15559 immunogen is purified recombinant fragment of human eNOS expressed in E. Coli. MA5-15559 detects eNOS which has a predicted molecular weight of approximately 133kDa.
eNOS Monoclonal antibody specifically detects eNOS in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
eNOS | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P29474 | |
NOS3 | |
Purified recombinant fragment of human eNOS expressed in E. Coli. | |
RUO | |
4846 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
6H2 | |
Unconjugated | |
NOS3 | |
2310065A03Rik; cNOS; Constitutive NOS; Ecnos; EC-NOS; endothelial nitric oxide synthase; endothelial nitric oxide synthase 3; endothelial nitric oxide synthase NOS3; endothelial NOS; eNos; e-NOS; nitric oxide synthase; nitric oxide synthase 3; nitric oxide synthase 3 (endothelial cell); nitric oxide synthase 3 transcript variant eNOS-delta20; nitric oxide synthase 3 transcript variant eNOS-delta20-21; nitric oxide synthase 3 transcript variant eNOS-delta21; nitric oxide synthase 3, endothelial cell; nitric oxide synthase, endothelial; NOS; NOS type III; Nos3; Nos-3; NOSIII; OTTHUMP00000213183 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |