missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Opsin 3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534038
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey (100%) Dog, Panda (95%) Marmoset, Rat, Elephant (89%) Mouse, Horse (84%).
Opsin 3 Polyclonal antibody specifically detects Opsin 3 in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).Specifications
Opsin 3 | |
Polyclonal | |
Unconjugated | |
Opn3 | |
ECPN; Encephalopsin; ERO; NMO-1; Opn3; opsin (encephalopsin); opsin 3; opsin 3 (encephalopsin, panopsin); opsin-3; Panopsin; PPP1R116; protein phosphatase 1, regulatory subunit 116 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23596 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9H1Y3 | |
Opn3 | |
Synthetic 19 amino acid peptide from C-terminus of human OPN3 / Encephalopsin. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |