missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nidogen G3 Domain Monoclonal Antibody (JF2)
Nidogen G3 Domain Monoclonal Antibody (JF2)
Brand: Invitrogen MA517021
This item is not returnable.
View return policy
Description
Specifications
| Nidogen G3 Domain | |
| Monoclonal | |
| 1 mg/mL | |
| 0.5M acetic acid, 2M tris with 0.1% sodium azide | |
| P10493, P14543 | |
| NID1 | |
| Recombinant fragment corresponding to the G3 region of mouse nidogen produced in Fisher rat plus YB2/0 cells. | |
| 50 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
| ELISA, Inhibition Assays, Western Blot | |
| JF2 | |
| Unconjugated | |
| NID1 | |
| A630025O17; enactin; Ent; Ent1; Entactin; entactin 1; entactin-1; NID; NID 1; Nid1; NID-1; Nidogen (enactin); nidogen 1; Nidogen precursor (Entactin); nidogen-1 | |
| Rat | |
| Protein G | |
| RUO | |
| 18073, 4811 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction