Learn More
EMX2, Mouse, Clone: 3H8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant EMX2.
Brand: Abnova H00002018-M07.100ug
Description
This gene encodes a homeobox-containing transcription factor that is the homolog to the 'empty spiracles' gene in Drosophila. Research on this gene in humans has focused on its expression in three tissues: dorsal telencephalon, olfactory neuroepithelium, and urogenetial system. It is expressed in the dorsal telencephalon during development in a low rostral-lateral to high caudal-medial gradient and is proposed to pattern the neocortex into defined functional areas. It is also expressed in embryonic and adult olfactory neuroepithelia where it complexes with eukaryotic translation initiation factor 4E (eIF4E) and possibly regulates mRNA transport or translation. In the developing urogenital system, it is expressed in epithelial tissues and is negatively regulated by HOXA10. Alternative splicing results in multiple transcript variants encoding distinct proteins
Sequence: HSPHPLFASQQRDPSTFYPWLIHRYRYLGHRFQGNDTSPESFLLHNALARKPKRIRTAFSPSQLLRLEHAFEKNHYVVGAERKQLAHSLSLTETQVKVSpecifications
| EMX2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_004098 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2018 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| ELISA | |
| 3H8 | |
| Mouse monoclonal antibody raised against a partial recombinant EMX2. | |
| EMX2 | |
| EMX2 | |
| EMX2 (NP_004089, 103 a.a. ∼ 200 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.