missing translation for 'onlineSavingsMsg'
Learn More

Elk-1 (pS383) Mouse, Alexa Fluor 647, Clone: M21-1721, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 560413

Product Code. 15860179

  • ONLINE EXCLUSIVE PRICE
    £301.00
    £289.48 / Each
    Save £11.52
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Elk-1 is a 62kDa member of the Ets oncogene family of transcription factors and a member of the subfamily of ternary complex factors (TCF). Ets proteins mediate a variety of gene activities in response to serum and growth factors. Proteins in the TCF subfamily form a ternary complex by binding to the Serum Response Element (SRE) in conjunction with a dimer of Serum Response Factors (SRF). Elk-1 is phosphorylated by mitogen-activated protein (MAP) kinase pathways in vivo at a cluster of S/T motifs at its carboxy-terminus.
Phosphorylation at these sites, particularly at serine 383 (S383), is critical for Elk-1 transcriptional activation. Studies have shown that Elk-1 is a direct target of activated MAP kinase, and that it is also a target of the stress-activated kinase SAPK/JNK. The M21-1721 monoclonal antibody recognizes the phosphorylated S383 of activated Elk-1. The orthologous phosphorylation sites of mouse and rat Elk-1 are S384 and S382, respectively.

Host Species: Mouse
Clone: M21-1721
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Phosphorylated Human Elk-1 Peptide

Intracellular Staining

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Elk-1 (pS383)
Monoclonal
Alexa Fluor 647
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Phosphorylated Human Elk-1 Peptide
50 Tests
Cell Biology
Human, Mouse, Rat
IgG1 κ
Flow Cytometry
M21-1721
ELK1
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.