Learn More
eukaryotic translation initiation factor 4 gamma, 1, Mouse, Clone: 2A9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant EIF4G1.
Brand: Abnova H00001981-M10.100ug
Description
The protein encoded by this gene is a component of the protein complex EIF4F, which is involved in the recognition of the mRNA cap, ATP-dependent unwinding of 5'-terminal secondary structure, and recruitment of mRNA to the ribosome. Alternative splicing results in five transcript variants encoding four distinct isoforms. [provided by RefSeq]
Sequence: DVAVLKARAKLLQKYLCDEQKELQALYALQALVVTLEQPPNLLRMFFDALYDEDVVKEDAFYSWESSKDPAEQQGKGVALKSVTAFFKWLREAEEESDHNSpecifications
| eukaryotic translation initiation factor 4 gamma, 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_182917 | |
| EIF4G1 | |
| EIF4G1 (NP_886553, 1500 a.a. ∼ 1599 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 2A9 | |
| Mouse monoclonal antibody raised against a partial recombinant EIF4G1. | |
| EIF4G1 | |
| DKFZp686A1451/EIF4F/EIF4G/p220 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 1981 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.