missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-EGFR, PLCG2 Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0560.1Set
Includes: EGFR rabbit purified polyclonal antibody (20μg); PLCG2 mouse purified polyclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the EGFR protein, and the other against the PLCG2 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
| EGFR | |
| Monoclonal/Polyclonal | |
| EGFR | |
| EGFR | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. EGFR rabbit purified polyclonal antibody (20 ug) 2. PLCG2 mouse purified polyclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
| Purified |
| In situ PLA | |
| Unconjugated | |
| ERBB/ERBB1/HER1/PIG61/mENA | |
| Mouse, Rabbit | |
| RUO | |
| 1956/5336 | |
| Human | |
| EGFR & PLCG2 Protein Protein Interaction Antibody Pair |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction