Learn More
epidermal growth factor receptor (erythroblastic leukemia viral (v-erb-b) oncogene homolog, avian), Mouse, Clone: 4H2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant EGFR.
Brand: Abnova H00001956-M02.100ug
Description
The protein encoded by this gene is a transmembrane glycoprotein that is a member of the protein kinase superfamily. This protein is a receptor for members of the epidermal growth factor family. EGFR is a cell surface protein that binds to epidermal growth factor. Binding of the protein to a ligand induces receptor dimerization and tyrosine autophosphorylation and leads to cell proliferation. Mutations in this gene are associated with lung cancer. [provided by RefSeq
Sequence: EEKKVCQGTSNKLTQLGTFEDHFLSLQRMFNNCEVVLGNLEITYVQRNYDLSFLKTIQEVAGYVLIALNTVERIPLENLQIIRGNMYYENSYALAVLSNYSpecifications
| epidermal growth factor receptor (erythroblastic leukemia viral (v-erb-b) oncogene homolog, avian) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_005228 | |
| EGFR | |
| EGFR (NP_005219, 26 a.a. ∼ 125 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Cell Cycle | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 4H2 | |
| Mouse monoclonal antibody raised against a partial recombinant EGFR. | |
| EGFR | |
| ERBB/ERBB1/HER1/PIG61/mENA | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 1956 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.