Learn More
EGF Receptor Mouse anti-Human, PE, Clone: EGFR1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555997
Description
Reacts with epidermal growth factor receptor (EGFR), a glycoprotein of approximately 170kDa, expressed on most cells. It consists of a glycosylated extracellular domain which recognizes epidermal growth factor (EGF), a transmembrane domain, and an intracellular domain with tyrosine-kinase activity essential for signal transduction. EGFR plays an important role in the growth and differentiation of many cellular types. Transforming growth factor (TGFα) and vaccinia virus growth factor also interact with EGFR.
Host Species: Mouse
Clone: EGFR.1
Isotype: IgG2b, κ
Species Reactivity: Human
Applications:
Flow Cytometry, Immunohistochemistry (Frozen)
Specifications
| EGF Receptor | |
| Monoclonal | |
| PE | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG2b κ |
| Flow Cytometry | |
| EGFR1 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.