Learn More
EGF Receptor (pY1173) Mouse, Unlabeled, Clone: 9H2, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 558382
Description
Epidermal Growth Factor (EGF) elicits a variety of cellular responses that are initiated by EGF Receptor (EGFR) binding and activation of intrinsic tyrosine kinase activity. EGFR, also known as ErbB1 or HER1, is a member of the ErbB class of receptor protein tyrosine kinases. It has an extracellular ligand-binding domain, a single transmembrane region, and a cytoplasmic region containing a protein tyrosine kinase domain and a c-terminal regulatory domain with many phosphorylation sites. Following ligand binding, EGFR forms homodimers and heterodimers with ErbB2. Specific C-terminal tyrosine residues are then autophosphorylated and, in turn, bind to adaptor proteins, kinases, or protein tyrosine phosphatases. Specifically, phosphorylated tyrosine 1173 (Y1173) interacts with Shc, SHP1, and PLCγ, which mediate downstream signaling cascades and negative feedback regulation of EGFR activation. Inappropriate expression or mutations of EGFR and/or deregulation of its signaling pathways are associated with many types of cancer, making EGFR a promising target for cancer therapies. The 9H2 monoclonal antibody recognizes the phosphorylated Y1173 in the regulatory domain of human EGFR.
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
| EGF Receptor (pY1173) | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Phosphorylated Human EGF Receptor Peptide | |
| 0.1 mg | |
| Cell Biology | |
| Canine, Human, Mouse, Rat | |
| IgG1 κ |
| Western Blot | |
| 9H2 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.