Learn More
EGF Receptor Mouse anti-Human, Brilliant Violet 510, Clone: EGFR.1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 563344
Description
The EGFR.1 monoclonal antibody specifically binds to the epidermal growth factor receptor (EGF-R). The EGF-R is a transmembrane glycoprotein ofapproximately 170 kDa that is expressed on most cells. It consists of a glycosylated extracellular domain which binds to epidermal growth factor (EGF), a transmembrane domain, and an intracellular domain with tyrosine-kinase activity essential for signal transduction. The EGF-R plays an important role in the growth and differentiation of many cellular types. Transforming growth factor α (TGFα), vaccinia virus growth factor, and related growth factors can also bind to and signal through the EGF-R.
Specifications
| EGF Receptor | |
| Monoclonal | |
| Brilliant Violet 510 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2b κ |
| Flow Cytometry | |
| EGFR.1 | |
| EGFR; ERBB; ERBB1; HER1; mENA; PIG61 | |
| Mouse | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.