Learn More
anti-EGF, Polyclonal, Abcam™
Brand: Abcam AB9695-100UG
Description
Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Human
Immunogen: Highly pure (>98%) recombinant hEGF (human Epidermal Growth Factor)
Applications:
ELISA, Immunohistochemistry (Paraffin), Western Blotting
Specifications
| EGF | |
| 0.5mg/mL | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| IgG |
| Polyclonal | |
| Unconjugated | |
| Highly pure (>98%) recombinant hEGF (human Epidermal Growth Factor) | |
| 100 μg | |
| Human |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.