Learn More
eukaryotic translation elongation factor 1 delta (guanine nucleotide exchange protein), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant EEF1D.
Brand: Abnova H00001936-A01.50uL
Description
This gene encodes a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit functions as guanine nucleotide exchange factor. It is reported that this subunit interacts with HIV-1 Tat, and thus it represses the translation of host-cell, but not HIV-1, mRNAs. Several alternatively spliced transcript variants encoding multiple isoforms have been found for this gene. [provided by RefSeq
Sequence: MRSGKASCTLETVWEDKHKYEEAERRFYEHEATQAAASAQQLPAEGPAMNGPGQDDPEDADEAEAPDGGSRRDPRKSQDSRKPLQKKRKRSSpecifications
| eukaryotic translation elongation factor 1 delta (guanine nucleotide exchange protein) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant EEF1D. | |
| EEF1D | |
| EF-1D/EF1D/FLJ20897/FP1047 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 1936 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_032378 | |
| EEF1D | |
| EEF1D (NP_115754, 1 a.a. ∼ 91 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.