Learn More
eukaryotic translation elongation factor 1 alpha 2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant EEF1A2.
Brand: Abnova H00001917-A01.50uL
Description
This gene encodes an isoform of the alpha subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This isoform (alpha 2) is expressed in brain, heart and skeletal muscle, and the other isoform (alpha 1) is expressed in brain, placenta, lung, liver, kidney, and pancreas. This gene may be critical in the development of ovarian cancer. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: HTAHIACKFAELKEKIDRRSGKKLEDNPKSLKSGDAAIVEMVPGKPMCVESFSQYPPLGRFAVRDMRQTVAVGVIKNVEKKSGGAGKVTKSAQKAQKAGSpecifications
| eukaryotic translation elongation factor 1 alpha 2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant EEF1A2. | |
| EEF1A2 | |
| EEF1AL/EF-1-alpha-2/EF1A/FLJ41696/HS1/STN/STNL | |
| Mouse | |
| 50 μL | |
| Yes | |
| 1917 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_001958 | |
| EEF1A2 | |
| EEF1A2 (NP_001949, 364 a.a. ∼ 462 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.