Learn More
EEA1 Mouse, Unlabeled, Clone: 14, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610457
Description
Early endosomes are a major cytoplasmic sorting compartment from which receptors and ligands may be distributed to various sites within the cell. Early endosome antigen 1 (EEA1) is a 180kDa hydrophilic peripheral membrane protein present in cytosol and membrane fractions. Immunofluorescence studies show that EEA1 colocalizes to early endosomes with transferrin receptor and Rab5, but not with the late endosome-localizing Rab7. EEA1 is predominantly α-helical and shares 17-20% sequence identity with the myosins. It contains a calmodulin-binding IQ motif and metal-binding cysteine “finger” motifs. It is thought that EEA1 is required for vesicular transport of proteins through early endosomes and that these finger motifs are required for this activity.
Host Species: Mouse
Clone: 14
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Mouse TRE1 aa. 82-179
Immunofluorescence, Immunoprecipitation
Specifications
| EEA1 | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| 150 μg | |
| Cell Biology | |
| Canine, Chicken, Human, Rat | |
| IgG1 |
| Western Blot | |
| 14 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.