Learn More
ectodysplasin A2 receptor, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human EDA2R protein.
Brand: Abnova H00060401-D01P.100ug
Description
EDA-A1 and EDA-A2 are two isoforms of ectodysplasin that are encoded by the anhidrotic ectodermal dysplasia (EDA) gene. Mutations in EDA give rise to a clinical syndrome characterized by loss of hair, sweat glands, and teeth. The protein encoded by this gene specifically binds to EDA-A2 isoform. This protein is a type III transmembrane protein of the TNFR (tumor necrosis factor receptor) superfamily, and contains 3 cysteine-rich repeats and a single transmembrane domain but lacks an N-terminal signal peptide. Multiple alternatively spliced transcript variants have been found for this gene, but some variants lack sufficient support. [provided by RefSeq
Sequence: MDCQENEYWDQWGRCVTCQRCGPGQELSKDCGYGEGGDAYCTACPPRRYKSSWGHHRCQSCITCAVINRVQKVNCTATSNAVCGDCLPRFYRKTRIGGLQDQECIPCTKQTPTSEVQCAFQLSLVEADAPTVPPQEATLVALVSSLLVVFTLAFLGLFFLYCKQFFNRHCQRGGLLQFEADKTAKEESLFPVPPSKETSAESQVSENIFQTQPLNPILEDDCSSTSGFPTQESFTMASCTSESHSHWVHSPIECTELDLQKFSSSASYTGAETLGGNTVESTGDRLELNVPFEVPSPSpecifications
| ectodysplasin A2 receptor | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human EDA2R protein. | |
| EDA2R | |
| EDA-A2R/EDAA2R/TNFRSF27/XEDAR | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_021783 | |
| EDA2R | |
| EDA2R (NP_068555.1, 1 a.a. ∼ 297 a.a) full-length human protein. | |
| 100 μg | |
| Membrane Receptors | |
| 60401 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.