missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EDA Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA542588
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
Peptide sequence: GPPGPPGPQG PPGLQGPSGA ADKAGTRENQ PAVVHLQGQG SAIQVKNDLS Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
EDA is a type II membrane protein that can be cleaved by furin to produce a secreted form. This protein, which belongs to the tumor necrosis factor family, acts as a homotrimer and may be involved in cell-cell signaling during the development of ectodermal organs. Defects in the gene for EDA are a cause of ectodermal dysplasia, anhidrotic, which is also known as X-linked hypohidrotic ectodermal dysplasia.
Specifica
| EDA | |
| Polyclonal | |
| Unconjugated | |
| EDA | |
| ECTD1; ectodermal dysplasia 1, anhidrotic; ectodermal dysplasia protein; Ectodysplasin; Ectodysplasin 1, isoform A2; ectodysplasin A; ectodysplasin-1; Ectodysplasin-A; Ectodysplasin-A, membrane form; Ectodysplasin-A, secreted form; Ed1; ED1-A1; ED1-A2; EDA; EDA 1; EDA protein; EDA protein homolog; EDA1; EDA2; Eda-A1; EDA-A2; HED; HED1; nackt; nkt; ODT1; oligodontia 1; RGD1563178; si:ch73-223d24.5; STHAGX1; Ta; tabby; Tabby protein; TNLG7C; tumor necrosis factor ligand 7C; XHED; XLHED; X-linked anhidroitic ectodermal dysplasia protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1896 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q92838 | |
| EDA | |
| Synthetic peptide directed towards the middle region of human EDA. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto