missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-EDA, Clone: 2F2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00001896-M03.100ug
This item is not returnable.
View return policy
Specifications
| EDA | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001399.1 | |
| EDA | |
| EDA (NP_001390.1 , 245 a.a. ∼ 391 a.a) partial recombinant protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2F2 | |
| Mouse monoclonal antibody raised against a partial recombinant EDA. | |
| EDA | |
| ED1/ED1-A1/ED1-A2/EDA1/EDA2/HED/XHED/XLHED | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1896 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction