Learn More
anti-E2F6, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00001876-PW1.1Set
Product Code. 16094748
  • £729.00 / 1 set
Includes: Antibody pair for IP: rabbit polyclonal anti-E2F6 (300μL), Antibody pair for WB: mouse polyclonal anti-E2F6 (50μL)
Specifications
E2F6
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
E2F-6/MGC111545
Mouse, Rabbit
RUO
1876
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
E2F6
E2F6
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-E2F6 (300 ul) 2. Antibody pair for WB: mouse polyclonal anti-E2F6 (50 ul)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.