missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E. coli serotype O/K Polyclonal Antibody, HRP

Goat Polyclonal Antibody
Brand: Invitrogen™ PA173034
This item is not returnable.
View return policy
Description
The PA1-73034 antibody reacts with E. coli. PA1-73034 has been successfully used in ELISA applications. The PA1-73034 antibody was raised against E. coli serotypes O157:H7, O20, O125, O55, O111 & K12.
Escherichia coli (E. coli) are Gram negative enteric bacteria that live in the human gut. Its presence in nature is an indication of human feacal pollution. E. coli also causes human disease such as urinary tract infections and neonatal meningitis. Infection with particular enterotoxigenic strains can lead to life threatening intestinal diseases. E. coli are distinguished immunologically by serotyping based on three types of antigen: the somatic or O antigen which corresponds to terminal sugars on the cell surface lipopolysaccharide, the capsular or K antigens and the flagellar or H antigens.
Specifications
| E. coli serotype O/K | |
| Polyclonal | |
| HRP | |
| Escherichia coli | |
| E. coli serotypes O157:H7, O20, O125, O55, O111 & K12. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA | |
| 1.0 mg/mL | |
| PBS with 10mg/mL BSA and 0.002% thimerosal | |
| Goat | |
| IgG fraction | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction