Learn More
E-Cadherin Mouse, Unlabeled, Clone: 36/E-CADHERIN, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610181
Description
E-Cadherin is a 120-kDa transmembrane glycoprotein that is localized in the adherens junctions of epithelial cells. There it interacts with the cytoskeleton through the associated cytoplasmic catenin proteins. In addition to being a calcium-dependent adhesion molecule, E-Cadherin is also a critical regulator of epithelial junction formation. Its association with catenins is necessary for cell-cell adhesion. These E-cadherin/catenin complexes associate with cortical actin bundles at both the zonula adherens and the lateral adhesion plaques. Tyrosine phosphorylation can disrupt these complexes, leading to changes in cell adhesion properties. E-Cadherin expression is often down-regulated in highly invasive, poorly differentiated carcinomas. Increased expression of E-Cadherin in these cells reduces invasiveness. Thus, loss of expression or function of E-Cadherin appears to be an important step in tumorigenic progression. The 36/E-Cadherin monoclonal antibody recognizes the cytoplasmic domain of E-Cadherin, regardless of phosphorylation status. The peptide immunogen was generated from human E-Cadherin aa. 735-883.
Note: Investigators are advised that this antibody has some degree of cross-reactivity to P-Cadherin.
Specifications
E-Cadherin | |
Monoclonal | |
250μg/mL | |
CD324; CDH1; CADH1; Cadherin-1; ECAD; CDHE; Arc-1; LCAM; UVO; Uvomorulin | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Western Blot | |
36/E-CADHERIN | |
Unconjugated | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG2a, κ | |
50 μg | |
Cell Biology | |
Canine, Human, Mouse, Rat | |
IgG2a κ |
For Research Use Only.