missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DYX1C1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518347
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
This gene encodes a tetratricopeptide repeat domain-containing protein. The encoded protein interacts with estrogen receptors and the heat shock proteins, Hsp70 and Hsp90. A chromosomal translocation involving this gene is associated with a susceptibility to developmental dyslexia. Mutations in this gene are associated with deficits in reading and spelling. Alternate splicing results in multiple transcript variants.
Specifications
| DYX1C1 | |
| Polyclonal | |
| Unconjugated | |
| DNAAF4 | |
| 1700010I24Rik; b2b811Clo; CILD25; DNAAF4; Dynein assembly factor 4, axonemal; dynein, axonemal, assembly factor 4; dyslexia susceptibility 1 candidate 1; dyslexia susceptibility 1 candidate 1 homolog; dyslexia susceptibility 1 candidate 1 homolog (human); dyslexia susceptibility 1 candidate gene 1 protein; Dyslexia susceptibility 1 candidate gene 1 protein homolog; DYX1; Dyx1c1; DYXC1; Edem2; Ekn1; FLJ37882; MGC70618; RD | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 161582 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8WXU2 | |
| DNAAF4 | |
| Synthetic peptide sequence (KIRNVIQGTELKS) corresponding to the C-terminus amino acids of DYX1C1 (aa 408-420). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction